Order Entry
Czech Republic
ContactUsLinkComponent
4-(Trifluoromethoxy)benzenesulphonyl chloride, Sigma-Aldrich®
4-(Trifluoromethoxy)benzenesulphonyl chloride, Sigma-Aldrich®
Catalog # SIAL386545-5G
Supplier:  Merck
CAS Number:  
4-(Trifluoromethoxy)benzenesulphonyl chloride, Sigma-Aldrich®
Catalog # SIAL386545-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :260.62 g/mol
Boiling Pt:120…122 °C (15 mmHg)
Density:1.536 g/cm³ (20 °C)
ADR:8,II
 :M-Clarity™ quality level = MQ100
EINECS:302-397-9
UN:3265
CAS Number:94108-56-2
Formula:C₇H₄ClF₃O₃S
MDL Number:MFCD00042408