1,3-Difluoro-4-nitrobenzene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:446-35-5
:
Formula:C₆H₃F₂NO₂
MDL Number:MFCD00007050
:159.09 g/mol
Boiling Pt:203 °C (1013 hPa)
Melting Pt:10 °C
Density:1.4571 g/cm³ (14 °C)
EINECS:207-167-0
ADR:6.1,III
:M-Clarity™ quality level = MQ100
UN:2810
Frequently Bought Together