


4-Fluorophenyl isocyanate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1195-45-5
:
Formula:C₇H₄FNO
MDL Number:MFCD00002023
:137.11 g/mol
Boiling Pt:42…47 °C (1 mmHg)
Density:1.206 g/cm³
Flash Pt:60 °C
EINECS:214-799-0
UN:3080
ADR:6.1,II
:GHS06
:GHS08
:GHS02
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5130 to 1.5150 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together