
Methyl-4-fluorobenzoate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:403-33-8
:
Formula:C₈H₇FO₂
MDL Number:MFCD00017959
:154.14 g/mol
Boiling Pt:195…197 °C
Density:1.192
Flash Pt:77 °C (170 °F)
EINECS:206-956-7
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together