Order Entry
Czech Republic
ContactUsLinkComponent
4-Fluorobenzoyl chloride 98%
Corrosive
4-Fluorobenzoyl chloride 98%
Certificates
undefined
Corrosive
4-Fluorobenzoyl chloride 98%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5299 to 1.5319 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together