Tetrafluoroterephthalonitrile, Sigma-Aldrich®
Catalog # SIAL104426-5G
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:5 g
- Sekvence:3173
Specifications
About this item
:
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:217-397-3
UN:3439
CAS Number:1835-49-0
Formula:C₈F₄N₂
MDL Number:MFCD00001776