
4-(Trifluoromethoxy)benzaldehyde 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:659-28-9
:
Formula:C₈H₅F₃O₂
MDL Number:MFCD00041530
:190.12 g/mol
Boiling Pt:80 °C (16 mmHg)
Density:1.33 g/cm³
Flash Pt:70 °C
EINECS:211-531-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow-green Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4570 to 1.4590 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together