Order Entry
Czech Republic
ContactUsLinkComponent
1,4-Difluorobenzene analytical standard, Supelco®
1,4-Difluorobenzene analytical standard, Supelco®
Catalog # 442249.
Supplier:  Merck
CAS Number:  
undefined
1,4-Difluorobenzene analytical standard, Supelco®
Catalog # 442249.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :114.09 g/mol
Merck Index:13,03172
Boiling Pt:88 °C (1013 hPa)
Melting Pt:–13 °C
Density:1.11 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-742-9
UN:1993
ADR:3,II
CAS Number:540-36-3
Formula:C₆H₄F₂
MDL Number:MFCD00000344