Potassium hydrogen phthalate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:877-24-7
:
Formula:HOOCC₆H₄COOK
MDL Number:MFCD00013070
:204.22 g/mol
Merck Index:13,07694
Melting Pt:295…300 °C
Density:1.6362 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:212-889-4
Frequently Bought Together