Potassium hydrogen phthalate, Certipur® Reag. Ph. Eur., Reag. USP volumetric standard, secondary reference material for alkalimetry, Supelco®
1.02400.0080
: Merck
:
- Pack type:Glass bottle
- Pk:80 g
- Seq:3392
- Solubility:80 g/l (20 °C)
- Melting Point:295 - 300 °C
- Molar Mass:204.22 g/mol
- Bulk Density:900 kg/m3
- Density:1.636 g/cm3
- pH:4.0 (50 g/l, H2O, 20 °C)
:877-24-7
:
:HOOCC₆H₄COOK
:MFCD00013070
:204.22 g/mol
:13,07694
:295…300 °C
:1.636 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:Ambient
:212-889-4