Order Entry
Czech Republic
ContactUsLinkComponent
Thiomersal 97.0-101.0%
Toxic
Health
Environment
Thiomersal 97.0-101.0%
Certificates
undefined
Toxic
Health
Environment
Thiomersal 97.0-101.0%
Certificates

Specification Test Results

  • Appearance:
    White to cream Powder
  • Infrared spectrum:
    Conforms
  • Assay:
    97.0 to 101.0 % (On dried substance)
  • Loss on drying:
    ≤0.5 % (vacuum/P2O5)
  • Additional info:
    ion mercuric: ≤ 0.7%
  • pH:
    6 to 8 (1 % solution)

Specification Test Results

Related Information

Frequently Bought Together