Benzoic acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:65-85-0
:
Formula:C₇H₆O₂
MDL Number:MFCD00002398
:122.12 g/mol
Merck Index:13,01092
Boiling Pt:249 °C (1013 hPa)
Melting Pt:121…123 °C
Density:1.316 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
Flash Pt:121 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-618-2
REACH:01-2119455536-33
:GHS07
Frequently Bought Together