
Methyl-4-aminobenzoate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:619-45-4
:
Formula:C₈H₉NO₂
MDL Number:MFCD00007891
:151.16 g/mol
Boiling Pt:299 °C (1013 hPa)
Melting Pt:110…114 °C
Density:1.216 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-598-7
:GHS07
Related Information
Frequently Bought Together