Order Entry
Czech Republic
ContactUsLinkComponent
Methyl-4-aminobenzoate 98%
Irritant
Methyl-4-aminobenzoate 98%
Certificates
undefined
Irritant
Methyl-4-aminobenzoate 98%

Specification Test Results

  • Appearance:
    White to beige Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    110 to 113 °C
  • Titration with HClO4:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together