Benzocaine (ethyl-4-aminobenzoate), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Ethyl 4-aminobenzoate, Aethoform
CAS Number:94-09-7
:
Formula:C₉H₁₁NO₂
MDL Number:MFCD00007892
:165.19 g/mol
Merck Index:13,01087
Boiling Pt:311 °C (1013 hPa)
Melting Pt:89…90 °C
:M-Clarity™ quality level = MQ200
Flash Pt:112 °C
EINECS:202-303-5
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together