Ethyl anthranilate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:87-25-2
:
Formula:C₉H₁₁NO₂
MDL Number:MFCD00007711
:165.19 g/mol
Boiling Pt:268 °C (1013 hPa)
Density:1.12 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
EINECS:201-735-1
Frequently Bought Together