Ethyl anthranilate, Sigma-Aldrich®
Catalog # SIAL242977-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:165.19 g/mol
Boiling Pt:268 °C (1013 hPa)
Density:1.12 g/cm³ (20 °C)
EINECS:201-735-1
CAS Number:87-25-2
Formula:C₉H₁₁NO₂
MDL Number:MFCD00007711