Ethyl-4-(dimethylamino)benzoate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10287-53-3
:
Formula:C₁₁H₁₅NO₂
MDL Number:MFCD00009115
:193.25 g/mol
Boiling Pt:112 °C (0.02 mmHg)
Melting Pt:63…65 °C
Density:1.0099 g/cm³ (99.7 °C)
:M-Clarity™ quality level = MQ200
EINECS:233-634-3
Frequently Bought Together