

Terephthaloyl dichloride 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-20-9
:
Formula:C₈H₄Cl₂O₂
MDL Number:MFCD00000693
:203.02 g/mol
Boiling Pt:266 °C (1013 hPa)
Melting Pt:79…83 °C
Density:1,34 g/cm³ (100 °C)
Flash Pt:180 °C
EINECS:202-829-5
UN:2923
ADR:8,I
:GHS05
:GHS06
Specification Test Results
- Appearance:White Crystals or crystalline flakes and/or chunks
- Infrared spectrum:Conforms
- Melting point:79 to 83 °C
- GC:≥99.0 %
Specification Test Results
Frequently Bought Together