

4-Anisoyl chloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-07-2
:
Formula:C₈H₇ClO₂
MDL Number:MFCD00000687
Merck Index:13,00676
Boiling Pt:145 °C (14 mmHg)
Melting Pt:22 °C
Density:1.26 g/cm³
Flash Pt:87 °C
UN:1729
ADR:8,II
:170.6 g/mol
EINECS:202-816-4
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to brown Liquid after melting
- Infrared spectrum:Conforms
- Refractive index:1.5800 to 1.5820 (20°C, 589 nm)
- GC:≥98.5 %
- HPLC:≥96.0 %
- Residue after evaporation:≤0.0005 %
- Free amine:≤0.001 %
Specification Test Results
Frequently Bought Together