

2-Anisoyl chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:21615-34-9
:
Formula:C₈H₇ClO₂
MDL Number:MFCD00000664
:170.6 g/mol
Boiling Pt:137 °C (1013 hPa)
Density:1.57 g/cm³ (20 °C)
EINECS:244-477-5
UN:1729
ADR:8,II
:M-Clarity™ quality level = MQ100
:GHS05
:GHS07
Frequently Bought Together