
4-Nitrobenzoyl chloride, Supelco®
Catalog # 73120-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
Merck Index:12,06687
Boiling Pt:278 °C (1013 hPa)
Melting Pt:97…98 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:M-Clarity™ quality level = MQ100
:185.57 g/mol
EINECS:204-517-4
:GHS05
CAS Number:122-04-3
Formula:C₇H₄ClNO₃
MDL Number:MFCD00007345