
4-Nitrobenzoyl chloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:122-04-3
:
Formula:C₇H₄ClNO₃
MDL Number:MFCD00007345
Merck Index:12,06687
Boiling Pt:202…205 °C (105 mmHg)
Melting Pt:70…74 °C
Flash Pt:102 °C
EINECS:204-517-4
ADR:8,II
:185.57 g/mol
UN:3261
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow Crystals or flakes
- Infrared spectrum:Conforms
- Melting point:70 to 74 °C
- Titration RCOCl:≥97.5 %
Specification Test Results
Frequently Bought Together