3-Nitrobenzoyl chloride, Sigma-Aldrich®
Catalog # SIAL127663-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:185.57 g/mol
Boiling Pt:275 °C (1013 hPa)
Melting Pt:30…33 °C
Density:1.42 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-505-9
UN:3261
ADR:8,II
CAS Number:121-90-4
Formula:C₇H₄ClNO₃
MDL Number:MFCD00007247