ContactUsLinkComponent
3,6-Dihydroxyphthalonitrile 98%
Toxic
3,6-Dihydroxyphthalonitrile 98%
Certificates
undefined
Toxic
3,6-Dihydroxyphthalonitrile 98%
Specification Test Results
  • Appearance:
    Brown to khaki Crystalline powder or needles
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥97.5 %