
2-Chloro-6-nitrophenol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:603-86-1
:
Formula:C₆H₄ClNO₃
MDL Number:MFCD00024233
:173.56 g/mol
Melting Pt:69…71 °C
Storage Temperature:Ambient
EINECS:210-061-7
UN:2811
ADR:6.1,III
:GHS07
Related Information
Frequently Bought Together