


Dicloran (2,6-dichloro-4-nitroaniline)
Catalog # EHERC12560000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:207.02 g/mol
Boiling Pt:323 °C (1013 hPa)
Melting Pt:186…189 °C
Storage Temperature:Ambient
EINECS:202-746-4
UN:2237
ADR:6.1,III
:GHS06
:GHS08
:GHS09
CAS Number:99-30-9
Formula:C₆H₄Cl₂N₂O₂
MDL Number:MFCD00007677