
2,6-Dichlorophenyl isothiocyanate, Sigma-Aldrich®
Catalog # SIAL475890-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:204.08 g/mol
Boiling Pt:100 °C (0.3 mmHg)
Melting Pt:44 °C
Storage Temperature:Refrigerator
ADR:8,III
:M-Clarity™ quality level = MQ100
UN:2923
:GHS05
CAS Number:6590-95-0
Formula:C₇H₃Cl₂NS
MDL Number:MFCD00041056