Order Entry
Czech Republic
ContactUsLinkComponent
 
 
3,5-Dichlorophenyl isocyanate 96%
 
undefined
 
3,5-Dichlorophenyl isocyanate 96%

 

  • Appearance:
    White to light yellow Low melting solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    29 to 35 °C
  • GC:
    ≥95 %

 

 

Frequently Bought Together