
3-Chloro-4-fluorobenzenesulphonyl chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:91170-93-3
:
Formula:C₆H₃Cl₂FO₂S
MDL Number:MFCD00041444
Boiling Pt:104…105 °C (1 mmHg)
Density:1.610
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
UN:3265
ADR:8,II
:229.06 g/mol
:GHS05
Related Information
Frequently Bought Together