4-Chloro-1,3-difluorobenzene, Sigma-Aldrich®
Catalog # SIAL335789-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:148.54 g/mol
Boiling Pt:127 °C (1013 hPa)
Melting Pt:–26 °C
Density:1.353 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
CAS Number:1435-44-5
Formula:C₆H₃ClF₂
MDL Number:MFCD00042569