
5-Amino-2-chlorobenzoic acid, Sigma-Aldrich®
Catalog # SIAL532495-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:171.58 g/mol
Boiling Pt:330 °C (1013 hPa)
Melting Pt:188 °C
Density:1.519 g/cm³ (15 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:201-916-5
:GHS07
CAS Number:89-54-3
Formula:C₇H₆ClNO₂
MDL Number:MFCD00007845