Methyl-4-chlorobenzoate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1126-46-1
:
Formula:C₈H₇ClO₂
MDL Number:MFCD00000621
:170.6 g/mol
Merck Index:14,2125
Boiling Pt:136…137 °C (10 mmHg)
Melting Pt:41…45 °C
Density:1.382
Flash Pt:106 °C (222 °F)
Storage Temperature:Ambient
EINECS:214-420-9
Related Information
Frequently Bought Together