Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
 

Ethyl 2-chlorobenzoate ≥98%

Ethyl 2-chlorobenzoate ≥98%
Certificates
Ethyl 2-chlorobenzoate ≥98%
Ethyl 2-chlorobenzoate ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 7335-25-3 Molecular Formula: C9H9ClO2 Formula Weight: 184.62 Flash Point: 110°(230°F) Refractive Index: 1.5240 Boiling Point: 243° Beilstein Reference: 2207359 MDL No.: MFCD00016338

Synonyms:  2-Chlorobenzoic acid ethyl ester
  • CAS Number:7335-25-3
  • Formula:C₉H₉ClO₂
  • MDL Number:MFCD00016338
  • Molecular Weight:184.62 g/mol
  • Boiling Pt:243 °C
  • Density:1.194
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:230-842-6
Product Family Options

Product Information