
1,3,5-Trichlorobenzene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:108-70-3
:
Formula:C₆H₃Cl₃
MDL Number:MFCD00000585
:181.45 g/mol
Merck Index:12,09761
Boiling Pt:208.4 °C (1013 hPa)
Melting Pt:63.4 °C
Density:1.4604 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:203-608-6
:GHS07
Storage Temperature:Ambient
Frequently Bought Together