
4-Bromobenzenesulphonyl chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:98-58-8
:
Formula:C₆H₄BrClO₂S
MDL Number:MFCD00007437
:255.52 g/mol
Merck Index:14,1407
Boiling Pt:152…154 °C (26 mmHg)
Melting Pt:73…77 °C
Density:1.37 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-683-2
UN:3261
ADR:8,II
:GHS05
Related Information
Frequently Bought Together