Order Entry
Czech Republic
ContactUsLinkComponent
 
 
2-Bromobenzenesulphonyl chloride 97%
 
undefined
 
2-Bromobenzenesulphonyl chloride 97%

 

  • Appearance:
    Light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    48 to 52 °C
  • Titration Argentometric:
    ≥96.0 % (ex Cl)

 

 

Frequently Bought Together