Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Registrovat
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromothioanisole
3-Bromothioanisole, Sigma-Aldrich®
3-Bromothioanisole, Sigma-Aldrich®
3-Bromothioanisole, Sigma-Aldrich®
SIAL549053-5G
:
Merck
:
3-Bromothioanisole, Sigma-Aldrich®
SIAL549053-5G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
203.1 g/mol
:
9,
:
M-Clarity™ quality level = MQ100
:
3334
:
33733-73-2
:
C₇H₇BrS
:
MFCD00041395