

4-Bromfenol, crystalline powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:106-41-2
:
Formula:C₆H₅BrO
MDL Number:MFCD00002313
Merck Index:13,01413
Boiling Pt:235…236 °C
Melting Pt:64…68 °C
Density:~ 1,84 g/cm³ (20 °C)
Flash Pt:≥ 110 °C
Storage Temperature:Ambient
EINECS:203-394-4
UN:2811
ADR:6.1,III
:GHS07
:GHS09
Frequently Bought Together