
2,6-Dibromophenol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:608-33-3
:
Formula:C₆H₄Br₂O
MDL Number:MFCD00002152
:251.91 g/mol
Boiling Pt:255 °C (1013 hPa)
Melting Pt:55…56 °C
Storage Temperature:Ambient
EINECS:210-161-0
UN:2811
ADR:6.1,III
:M-Clarity™ quality level = MQ100
:GHS07
Frequently Bought Together