Order Entry
Czech Republic
ContactUsLinkComponent
2,6-Dibromo-4-nitroaniline, Sigma-Aldrich®
Irritant
2,6-Dibromo-4-nitroaniline, Sigma-Aldrich®
Catalog # SIAL190268-100G
Supplier:  Merck
CAS Number:  
Irritant
2,6-Dibromo-4-nitroaniline, Sigma-Aldrich®
Catalog # SIAL190268-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :295.92 g/mol
Melting Pt:204…209 °C
 :M-Clarity™ quality level = MQ100
EINECS:212-577-8
 :GHS07
Storage Temperature:Ambient
CAS Number:827-94-1
Formula:C₆H₄Br₂N₂O₂
MDL Number:MFCD00007639