3-Bromophenyl isothiocyanate, Sigma-Aldrich®
Catalog # SIAL253162-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:214.09 g/mol
Boiling Pt:256 °C (1013 hPa)
ADR:9,
:M-Clarity™ quality level = MQ100
UN:3335
CAS Number:2131-59-1
Formula:C₇H₄BrNS
MDL Number:MFCD00004803