Order Entry
Czech Republic
ContactUsLinkComponent
4-Bromophenyl isocyanate 99%
Health
Irritant
4-Bromophenyl isocyanate 99%
Catalog # ACRO221130100
CAS Number:  
undefined
Health
Irritant
4-Bromophenyl isocyanate 99%
Catalog # ACRO221130100
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    White to light beige Crystalline solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    39 to 42 °C
  • GC:
    ≥98.5 %

Specification Test Results

About this item

 :
 :198.02 g/mol
Merck Index:13,01415
Boiling Pt:158 °C (14 mmHg)
Melting Pt:42…44 °C
Flash Pt:109 °C
Storage Temperature:Refrigerator
EINECS:219-662-9
UN:3335
ADR:9,III
 :GHS08
 :GHS07
CAS Number:2493-02-9
Formula:C₇H₄BrNO
MDL Number:MFCD00002022