Order Entry
Czech Republic
ContactUsLinkComponent
3-Bromophenyl isocyanate 97%
Health
3-Bromophenyl isocyanate 97%
Certificates
undefined
Health
3-Bromophenyl isocyanate 97%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5835 to 1.5865 (20°C, 589 nm)
  • GC:
    ≥96.0 %

Specification Test Results

Related Information

Frequently Bought Together