
2-Bromo-5-fluorophenol, Sigma-Aldrich®
Catalog # SIAL380350-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
Boiling Pt:184…186 °C (1013 hPa)
Density:1.717 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:191 g/mol
:GHS07
CAS Number:147460-41-1
Formula:C₆H₄BrFO
MDL Number:MFCD00040939