0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
4-Bromo-2,6-difluoroaniline
4-Bromo-2,6-difluoroaniline, Sigma-Aldrich®
4-Bromo-2,6-difluoroaniline, Sigma-Aldrich®
SIAL327867-5G
:
Merck
:
4-Bromo-2,6-difluoroaniline, Sigma-Aldrich®
SIAL327867-5G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
208.01 g/mol
:
M-Clarity™ quality level = MQ200
:
67567-26-4
:
C₆H₄BrF₂N
:
MFCD00013389