Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Dibromobenzoic acid
3,5-Dibromobenzoic acid, Sigma-Aldrich®
3,5-Dibromobenzoic acid, Sigma-Aldrich®
SIAL411523-5G
:
Merck
:
3,5-Dibromobenzoic acid, Sigma-Aldrich®
SIAL411523-5G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
279.92 g/mol
:
M-Clarity™ quality level = MQ100
:
GHS07
:
Ambient
:
618-58-6
:
C₇H₄Br₂O₂
:
MFCD00051758