ContactUsLinkComponent
Methyl-3-bromobenzoate 98%
Irritant
Methyl-3-bromobenzoate 98%
Certificates
undefined
Irritant
Methyl-3-bromobenzoate 98%
Specification Test Results
  • Appearance:
    White to light yellow Low melting crystalline solid or liquid
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥97.5 %