Ethyl-3-bromobenzoate, Sigma-Aldrich®
SIAL335819-25G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
:
:229.07 g/mol
:259 °C (1013 hPa)
:1.4308 g/cm³ (18.9 °C)
:M-Clarity™ quality level = MQ100
:246-223-9
:24398-88-7
:C₉H₉BrO₂
:MFCD00013529