Ethyl-3-bromobenzoate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:24398-88-7
:
Formula:C₉H₉BrO₂
MDL Number:MFCD00013529
:229.07 g/mol
Boiling Pt:261 °C
Density:1.430
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:246-223-9
Related Information
Frequently Bought Together